Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
This peptide corresponds to 17 amino acid peptide near the amino terminus of human DDX41.
PEP-1593 can be used as a blocking peptide with polyclonal antibody PA5-34550. For use as a blocking peptide in Western blot, incubate the peptide with equal volume of antibody for 30 min at 37ºC.
Probable ATP-dependent RNA helicase DDX41 is a DEAD box protein characterized by the conserved motif Asp-Glu-Ala-Asp. It is required during post-transcriptional gene expression and may be involved in pre-mRNA splicing. Mutations affecting the gene can result in myeloproliferative/lymphoproliferative neoplasms, familial (MPLPF).
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 2900024F02Rik; Abstrakt; DEAD (Asp-Glu-Ala-Asp) box polypeptide 41; DEAD box protein 41; DEAD box protein abstrakt homolog; DEAD-box protein abstrakt; Probable ATP-dependent RNA helicase DDX41; putative RNA helicase
Gene Aliases: ABS; DDX41; MPLPF
UniProt ID: (Human) Q9UJV9
Entrez Gene ID: (Human) 51428
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support