Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
For maximum recovery please spin prior to use. Unless noted below, aliquots of the 5 µg, 10 µg and 20 µg sizes of kinase are not recommended as materials can be used in original packaging until exhausted. For larger sizes, the number of freeze/thaws may be reduced by preparing aliquots, aliquots below 20 µL are not recommended. Please never store a kinase diluted. If properly stored at -80C, this product is guaranteed for 6 months from date of purchase.
This KSR2 recombinant human protein (554-end) was expressed in insect cells.
KSR2 recombinant human protein is also available in our SelectScreen™ Kinase Profiling Service.
• Validated for use with the Z'-LYTE™ assay
• Available in multiple pack sizes for convenient use
• Also available in our SelectScreen Kinase Profiling Service
Protein Form: Recombinant, Active, Catalytic
KSR2 serine/threonine kinase or kinase suppressor of Ras 2 specifically regulates the activity of MEKK3 and COT but not other members of the MAP3K family. KSR interacts with a regulatory Raf molecule in cis to induce a conformational switch of MEK, facilitating MEK's phosphorylation by a separate catalytic Raf molecule in trans. Blocks MAP3K8 kinase activity and MAP3K8- mediated signaling. Acts as a negative regulator of MAP3K3- mediated activation of ERK, JNK and NF-kappa-B pathways, inhibiting MAP3K3-mediated interleukin-8 production.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: hKSR2; Kinase suppressor of Ras 2
Gene Aliases: KSR2
UniProt ID: (Human) Q6VAB6
Entrez Gene ID: (Human) 283455
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support