Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
It is recommended that the protein be aliquoted for optimal storage.
It is recommended that sterile water (1099 µL) be added to the vial to prepare a stock solution of 0.91 mg/mL.
Concentration is measured by UV.
May play a role in triggering innate immune responses. Does not seem to play a role for any class I MHC antigen recognition.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CD85 antigen-like family member F; CD85f; CD85f antigen; ILT-11; Immunoglobulin-like transcript 11; immunoglobulin-like transcript 11 protein; leucocyte Ig-like receptor A5; leukocyte Ig-like receptor 9; Leukocyte immunoglobulin-like receptor 9; Leukocyte immunoglobulin-like receptor subfamily A member 5; leukocyte immunoglobulin-like receptor subfamily A member 5 soluble; leukocyte immunoglobulin-like receptor, subfamily A (with TM domain), member 5; leukocyte immunoglobulin-like receptor, subfamily B (with TM and ITIM domains), member 7; LIR-9
Gene Aliases: CD85; CD85F; ILT-11; ILT11; LILRA5; LILRB7; LIR-9; LIR9
UniProt ID: (Human) A6NI73
Entrez Gene ID: (Human) 353514
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support