Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
PEP-015 is an 11 amino acid (amino acids 29-39) synthetic peptide whose sequence is from the amino end of SERCA3 that is conserved in mouse, rat, and human SERCA3. The sequence of this peptide is (amino to carboxy terminus): V-T-D-A-R-E-R-Y-G-P-N
This peptide may be used for neutralization and control experiments with the polyclonal antibody that reacts with this product and serca3, catalog # PA1-910A. Using a solution with equal weights per unit volume of peptide and corresponding antibody will yield a solution with a large molar excess of peptide that is able to competitively bind the antibody.
Reconstitute with 0.1 mL of distilled water.
ATP dependent calcium pumps are responsible in part for the maintenance of low cytoplasmic free calcium concentrations. The ATP pumps that reside in intracellular organelles are encoded by a family of structurally related enzymes, termed the sarcoplasmic or endoplasmic reticulum calcium ATPases (SERCA). The SERCA2 gene is subject to tissue dependent processing which is responsible for the generation of SERCA2a muscle-specific isoform expressed in type I (slow) skeletal, cardiac and smooth muscle and the SERCA2b isoform expressed in all cell types. SERCA3 has been found to co-express with SERCA2b in platelets, mast cells, lymphoid cells, and epithelial cells. Due to this pattern of expression it is referred to as the non-muscle form of SERCA.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: adenosine triphosphatase, calcium; ATPase, Ca(2+)-transporting, ubiquitous; ATPase, Ca++ transporting, ubiquitous; Calcium pump 3; calcium-translocating P-type ATPase; sarco/endoplasmic reticulum Ca2+ -ATPase; sarcoendoplasmic reticulum Ca2+ ATPase type 3; Sarcoplasmic/endoplasmic reticulum calcium ATPase 3; SERCA3; SR Ca(2+)-ATPase 3
Gene Aliases: ATP2A3; SERCA3; SERCA3b
UniProt ID: (Human) Q93084, (Rat) P18596, (Mouse) Q64518
Entrez Gene ID: (Human) 489, (Rat) 25391, (Mouse) 53313
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support