Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
This peptide corresponds to 13 amino acids near the center of human NALP4.
PEP-1134 can be used as a blocking peptide with polyclonal antibody PA5-21020.
NALP proteins include the apoptosis regulator APAF1 (apoptotic protease activating factor 1) and mammalian NOD-LRR proteins and are thought to be involved in inflammation and reproduction. NALP4, also known as PYRIN-containing APAF1-like protein 4, has a C-terminal leucine-rich repeat (LRR) region, an N-terminal Pyrin domain (PYD) followed by a NACHT domain, and a NACHT-associated domain. In transfected 293 cells, NALP4 suppressed NF-kappa-B induction by TNF-a and IL-1-b, suggesting NALP4 operates at a point of convergence in these two signaling pathways. NALP4 also suppressed NF-kappa-B induction resulting from overexpression of several adapter proteins and protein kinases involved in these pathways, including TRAF2, TRAF6, RIP, and IRAK2, as well as the IKK-alpha and IKK-beta, suggesting that NALP4 is critical in modulating NF-kappa-B activity.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Cancer/testis antigen 58; CT58; NACHT, leucine rich repeat and PYD containing 4; NACHT, LRR and PYD containing protein 4; NACHT, LRR and PYD domains-containing protein 4; NLR family, pyrin domain containing 4; nucleotide-binding oligomerization domain, leucine rich repeat and pyrin domain containing 4; PAAD and NACHT-containing protein 2; PAN2; PYPAF4; PYRIN and NACHT-containing protein 2; PYRIN-containing APAF1-like protein 4; Ribonuclease inhibitor 2
Gene Aliases: CLR19.5; CT58; NALP4; NLRP4; PAN2; PYPAF4; RNH2
UniProt ID: (Human) Q96MN2
Entrez Gene ID: (Human) 147945
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support