Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
For maximum recovery please spin prior to use. Unless noted below, aliquots of the 5 µg, 10 µg and 20 µg sizes of kinase are not recommended as materials can be used in original packaging until exhausted. For larger sizes, the number of freeze/thaws may be reduced by preparing aliquots, aliquots below 20 µL are not recommended. Please never store a kinase diluted. If properly stored at -80C, this product is guaranteed for 6 months from date of purchase.
This NEK8 recombinant human protein (1-322) was expressed in insect cells.
NEK8 recombinant human protein is also available in our SelectScreen™ Kinase Profiling Service.
• Validated for use with the LanthaScreen™ Eu Kinase Binding assay
• Available in multiple pack sizes for convenient use
• Also available in our SelectScreen Kinase Profiling Service
Protein Form: Recombinant, Active, Catalytic
NEK8 serine/threonine kinase or NIMA-related kinase 8 is related to NIMA (never in mitosis, gene A) of Aspergillus nodulins and may play a role in cell cycle progression from G2 to M phase. NEK8 is part of a complex of ciliary proteins required for renal and cardiovascular development. NEK8 interacts with polycystin signal transduction pathways and may control the targeting of these ciliary proteins. Required for renal tubular integrity. May regulate local cytoskeletal structure in kidney tubule epithelial cells. May regulate ciliary biogenesis through targeting of proteins to the cilia.Defects in NEK8 are the cause of nephronophthisis type 9 (NPHP9) [MIM:613824]. NPHP9 is an autosomal recessive disorder resulting in end-stage renal disease. It is a progressive tubulo-interstitial kidney disorder histologically characterized by modifications of the tubules with thickening of the basement membrane, interstitial fibrosis and, in the advanced stages, medullary cysts.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: NEK 8; nephrocystin 9; Never in mitosis A-related kinase 8; NIMA (never in mitosis gene a)- related kinase 8; NIMA-family kinase NEK8; NIMA-related kinase 12a; NIMA-related kinase 8; Nima-related protein kinase 12a; NimA-related protein kinase 8; Serine/threonine-protein kinase Nek8; serine/thrionine-protein kinase NEK8
Gene Aliases: JCK; NEK12A; NEK8; NPHP9; RHPD2
UniProt ID: (Human) Q86SG6
Entrez Gene ID: (Human) 284086
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support