Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
This peptide corresponds to 17 amino acid peptide near the center of human RIM2.
PEP-1410 can be used as a blocking peptide with polyclonal antibody PA5-34367.
Rab3-interacting molecules (RIMs) are synaptic proteins necessary for neuronal transmission and plasticity. Rim1 and Rim2 proteins are expressed in overlapping but distinct patterns throughout the brain. While the ablation of either gene was not lethal in mice, the deletion of both resulted in postnatal mortality. This lethality is due to a defect in neurotransmitter release; synapses without RIM proteins can still release neurotransmitters but are unable to do so in response to normal Ca2+ triggers. Like Rim1, Rim2 is thought to be an effector protein for Rab3, binding to Rab3 on synaptic vesicles in a GTP-dependent manner.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: non-small cell lung cancer RimL3a protein; non-small cell lung cancer RimL3c protein; nuclear protein; Rab-3-interacting molecule 2; Rab-3-interacting protein 3; RAB3 interacting protein 3; rab3-interacting molecule 2; Rab3-interacting protein; Regulating synaptic membrane exocytosis protein 2; RIM 2; RIM2 (CT)
Gene Aliases: KIAA0751; OBOE; RAB3IP3; RIM2; RIMS2
UniProt ID: (Human) Q9UQ26
Entrez Gene ID: (Human) 9699
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support