Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
This peptide corresponds to 19 amino acid peptide near the carboxy terminus of human ZFX.
PEP-1419 can be used as a blocking peptide with polyclonal antibody PA5-34376.
ZFX is a member of the Kruppel C2H2-type zinc-finger protein family. The full-length protein contains an acidic transcriptional activation domain (AD), a nuclear localization sequence (NLS) and a DNA binding domain (DBD) consisting of 13 C2H2-type zinc fingers. Studies in mouse embryonic and adult hematopoietic stem cells showed that this gene was required as a transcriptional regulator for self-renewal of both stem cell types, but it was dispensable for growth and differentiation of their progeny.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: X-linked zinc finger protein; Zinc finger X-chromosomal protein
Gene Aliases: ZFX; ZNF926
UniProt ID: (Human) P17010
Entrez Gene ID: (Human) 7543
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support