Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Recombinant Murine Prokineticin-2 is an 11.6 kDa protein consisting of 103 amino acid residues, including ten cysteine residues that can potentially form five pairs of intra-molecular disulfide bonds.
This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis
Prokineticin-2 (PK2) is a cysteine-rich secreted protein that is expressed in the suprachiasmatic nucleus (SCN) with receptors located in the critical autonomic control centers of the brain. It has a depolarizing effect on neurons expressing the receptor. PK2 is predominantly controlled by the endogenous circadian clock, but light also plays a modulatory role. PK2 functions as a critical SCN output molecule responsible for circadian locomotor rhythms. PK2 expression is high during the day, and responsive to nocturnal light pulses. PK2 also functions as a chemoattractant for subventricular zone-derived neuronal progenitors.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Bombina variegata 8 kD protein; Bombina variegata 8kDa protein; Bv 8 homolog (Bombina variegata); PK2; prokineticin 1; Prokineticin-2; Protein Bv8 homolog
Gene Aliases: Bv8; PK2; Prok1; Prok2
UniProt ID: (Mouse) Q9QXU7
Entrez Gene ID: (Mouse) 50501
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support