Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
This peptide corresponds to 17 amino acids near the center of mouse SLPI.
PEP-0503 can be used as a blocking peptide with polyclonal antibody PA5-20386.
SLPI is a secreted inhibitor which protects epithelial tissues from serine proteases. It is found in various secretions including seminal plasma, cervical mucus, and bronchial secretions, and has affinity for trypsin, leukocyte elastase, and cathepsin G. Its inhibitory effect contributes to the immune response by protecting epithelial surfaces from attack by endogenous proteolytic enzymes; the protein is also thought to have broad-spectrum anti-biotic activity.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: ALP; Antileukoproteinase; RP23-462O16.7; Secretory leukocyte protease inhibitor
Gene Aliases: Slpi
UniProt ID: (Mouse) P97430
Entrez Gene ID: (Mouse) 20568
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support