Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
It is recommended that the protein be aliquoted for optimal storage.
It is recommended that sterile water (800 µL) be added to the vial to prepare a stock solution of 1.25 mg/mL.
Concentration is measured by UV.
T cell activation by APCs is positively and negatively regulated by members of the B7 family. VSIG4 is a strong negative regulator of murine and human T cell proliferation and IL-2 production.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: complement receptor of the immunoglobulin superfamily; V-set and immunoglobulin domain-containing protein 4
Gene Aliases: A530061A11; BC025105; CRIg; Z39IG
Entrez Gene ID: (Mouse) 278180
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support