Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Recombinant Rat Carboxypeptidase-B is a 35.1 kDa protein consisting of 307 amino acids.
This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis
Three different procarboxypeptidases A and two different procarboxypeptidases B have been isolated. The B1 and B2 forms differ from each other mainly in isoelectric point. Carboxypeptidase B1 is a highly tissue-specific protein and is a useful serum marker for acute pancreatitis and dysfunction of pancreatic transplants. It is not elevated in pancreatic carcinoma.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: carboxypeptidase B; pancreatic carboxypeptidase B1
Gene Aliases: Cpb
Entrez Gene ID: (Rat) 24271
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support