Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
ChIP-on-chip (location analysis) is a powerful microarray-based method for analyzing epigenetic modifications and DNA sequences bound to specific regulatory proteins. Briefly, in ChIP-on-chip protein-DNA complexes are crosslinked, immunoprecipitated, purified, amplified and labeled and then allowed to hybridize to a variety of high resolution arrays. Invitrogen has developed cost-effective products to overcome challenges facing ChIP microarray researchers – notably higher yields without increase non specific binding and more representative amplification.
Select your ChIP-on-chip array to view reagents appropriate for your research workflow:
Literature
Chromatin Research Topics
Tools
Primary Antibody Search
Questions
Send your questions to our top researchers at techline@lifetech.com.