Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
66061-1-Ig recognizes 14-3-3 tau (YWHAQ), eta (YWHAH), and zeta (YWHAZ), but not epsilon (YWHAE).
Immunogen sequence: MEKTELIQK AKLAEQAERY DDMATCMKAV TEQGAELSNE ERNLLSVAYK NVVGGRRSAW RVISSIEQKT DTSDKKLQLI KDYREKVESE LRSICTTVLE LLDKYLIANA TNPESKVFYL KMKGDYFRYL AEVACGDDRK QTIDNSQGAY QEAFDISKKE MQPTHPIRLG LALNFSVFYY EILNNPELAC TLAKTAFDEA IAELDTLNED SYKDSTLIMQ LLRDNLTLWT SDSAGEECDA AEGAEN (1-245 aa encoded by B C056867)
This gene product belongs to the 14-3-3 family of proteins which mediate signal transduction by binding to phosphoserine-containing proteins. This highly conserved protein family is found in both plants and mammals, and this protein is 99% identical to the mouse and rat orthologs. This gene is upregulated in patients with amyotrophic lateral sclerosis. It contains in its 5' UTR a 6 bp tandem repeat sequence which is polymorphic, however, there is no correlation between the repeat number and the disease.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 14-3-3 protein T-cell; 14-3-3 protein tau; 14-3-3 protein theta; 14-3-3 tau; 14-3-3 theta; OTTHUMP00000165887; Protein HS1; protein tau; protein, theta; tyrosine 3-monooxygenase/tryptophan 5-monooxygenase activation protein, theta polypeptide
Gene Aliases: 14-3-3; 14-3-3t; 1C5; 2700028P07Rik; AA409740; AU021156; HS1; R74690; YWHAQ
UniProt ID: (Human) P27348, (Rat) P68255, (Mouse) P68254
Entrez Gene ID: (Pig) 100048964, (Human) 10971, (Rat) 25577, (Mouse) 22630
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support