Search Thermo Fisher Scientific
Active Motif
FIGURE: 1 / 1
3-methylcytosine arises by erroneous methylation of DNA through spontaneous exposure to endogenous S-adenosyl methionine (SAM). Since the N3-position in cytosine is protected in double stranded DNA, this phenomenon mainly occurs in single stranded DNA. The resulting 3-mC is toxic and mutagenic and increased levels of 3-mC seem to impair cell proliferation. Therefore, it has to be repaired by the cell. This is established by oxidative demethylation, which is catalyzed by the AlkB protein.
Protein Aliases: 3-mC; 3-meC; 3-Methyl-cytosine; 3mc; C5H7NO3