Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: TQGSHLGSCI EKARPYYEAR RLAKEAQQET QKAALRYERA VSMHNAAREM VFVAEQGVMA DKNRLDPTWQ EMLNHATCK
Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 100%.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: SH3 domain-binding protein 5-like; SH3-binding domain protein 5 like; SH3-binding domain protein 5-like; SH3BP-5-like
Gene Aliases: KIAA1720; SH3BP5L; UNQ2766/PRO7133
UniProt ID: (Human) Q7L8J4
Entrez Gene ID: (Human) 80851
Molecular Function:
scaffold/adaptor protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support