Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
This target displays homology in the following species: Cow: 86%; Dog: 86%; Horse: 92%; Human: 100%; Mouse: 100%; Pig: 92%; Rabbit: 100%; Rat: 100%; Yeast: 100%; Zebrafish: 100%
4930567H17Rik is a protein coding gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: uncharacterized protein LOC619303
Entrez Gene ID: (Mouse) 619303
Molecular Function:
RNA metabolism protein
nucleic acid metabolism protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support