Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MPSYTVTVA TGSQWFAGTD DYIYLSLVGS AGCSEKHLLD KPFYNDFERG AVDSYDVTVD EELGEIQLVR IEKRKYWLND DWYLKYITLK TPHGDYIEFP CYRWITGDVE VVLRDGRAKL ARDDQIHILK QHRRKELETR QKQYRWMEWN PGFPLSIDAK CHKDLPRDIQ FDSEKGVDFV LNYSKAMENL FINRFMHMFQ SSWNDFADFE KIFVKISNTI SERVMNHWQE DLMFGYQFLN GCNPVLIRRC TELPEKLPVT TEMVECSLER QLSLEQEVQQ GNIFIVDFEL LDGIDANKTD PCTLQFLAAP ICLLYKNLAN KIVPIAIQLN QIPGDENPIF LPSDAKYDWL (1-349 aa encoded by BC130332)
This gene encodes a protein which, with Arachidonate 5-lipoxygenase-activating protein, is required for leukotriene synthesis. Leukotrienes are arachidonic acid metabolites which have been implicated in various types of inflammatory responses, including asthma, arthritis and psoriasis. This protein localizes to the plasma membrane. Inhibitors of its function impede translocation of 5-lipoxygenase from the cytoplasm to the cell membrane and inhibit 5-lipoxygenase activation.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 5-LO; Arachidonate 5-lipoxygenase; arachidonic 5-lipoxygenase alpha-10 isoform; arachidonic 5-lipoxygenase delta-10-13 isoform; arachidonic 5-lipoxygenase delta-13 isoform; arachidonic 5-lipoxygenase delta-p10 isoform; arachidonic acid 5-lipoxygenase; leukotriene A4 synthase; LOX-5; lox5; Polyunsaturated fatty acid 5-lipoxygenase; RP11-67C2.3
Gene Aliases: 5-LO; 5-LOX; 5LPG; ALOX5; LOG5
UniProt ID: (Human) P09917
Entrez Gene ID: (Pig) 100156205, (Human) 240
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support