Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
In direct ELISAs, 100% cross-reactivity with recombinant mouse 5T4 is observed.
Reconstitute at 0.5 mg/mL in sterile PBS.
This gene encodes a leucine-rich transmembrane glycoprotein that may be involved in cell adhesion. The encoded protein is an oncofetal antigen that is specific to trophoblast cells. In adults this protein is highly expressed in many tumor cells and is associated with poor clinical outcome in numerous cancers. Alternate splicing in the 5' UTR results in multiple transcript variants that encode the same protein. [provided by RefSeq, Oct 2009].
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 5T4 oncofetal antigen; 5T4 oncofetal trophoblast glycoprotein; 5T4 oncotrophoblast glycoprotein; 5T4-AG; M6P1; Trophoblast glycoprotein; WAIF1; Wnt-activated inhibitory factor 1
Gene Aliases: 5T4; 5T4AG; M6P1; TPBG; WAIF1
UniProt ID: (Human) Q13641
Entrez Gene ID: (Human) 7162
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support