Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Cell lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MEAVCLGTCP EPEASMSTAI PGLKKGNNAL QSIILQTLLE KENGQRKYGG PPPGWDAAPP ERGCEIFIGK LPRDLFEDEL IPLCEKIGKI YEMRMMMDFN GNNRGYAFVT FSNKVEAKNA IKQLNNYEIR NGRLLGVCAS VDNCRLFVGG IPKTKKREEI LSEMKKVTEG VVDVIVYPSA ADKTKNRGFA FVEYESHRAA AMARRKLLPG RIQLWGHGIA VDWAEPEVEV DEDTMSSVKI LYVRNLMLST SEEMIEKEFN NIKPGAVERV KKIRDYAFVH FSNREDAVEA MKALNGKVLD GSPIEVTLAK PVDKDSYVRY TRGTGGRGTM LQGEYTYSLG QVYDPTTTYL GAPVFYAPQT YAAIPSLHFP ATKGHLSNRA IIRAPSVRGA AGVRGLGGRG YLAYTGLGRG YQVKGDKRED KLYDILPGME LTPMNPVTLK PQGIKLAPQI LEEICQKNNW GQPVYQLHSA IGQDQRQLFL YKITIPALAS QNPAIHPFTP PKLSAFVDEA KTYAAEYTLQ TLGIPTDGGD GTMATAAAAA TAFPGYAVPN ATAPVSAAQL KQAVTLGQDL AAYTTYEVYP TFAVTARGDG YGTF
Mammalian apolipoprotein B mRNA undergoes site-specific Cto U deamination, which is mediated by a multi-component enzyme complex containing a minimal core composed of APOBEC-1 and acomplementation factor encoded by this gene. The gene product has three non-identical RNA recognition motifs and belongs to the hnRNPR family of RNA-binding proteins. It has been proposed that this complementation factor functions as an RNA-binding subunit and docks APOBEC-1 to deaminate the upstream cytidine. Studies suggest that the protein may also be involved in other RNA editing or RNA processing events.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: apo-B RNA editing protein; apobec-1 complementation factor (ACF) (ASP); APOBEC-1 stimulating protein; APOBEC1 complementation factor; APOBEC1-stimulating protein; MGC163391; RP11-564C4.2
Gene Aliases: A1CF; ACF; ACF64; ACF65; APOBEC1CF; ASP
UniProt ID: (Human) Q9NQ94
Entrez Gene ID: (Human) 29974
Molecular Function:
RNA metabolism protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support