Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ADORA2A is a protein which is one of several receptor subtypes for adenosine. The activity of the encoded protein, a G-protein coupled receptor family member, is mediated by G proteins which activate adenylyl cyclase. This protein is abundant in basal ganglia, vasculature and platelets and it is a major target of caffeine.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: A2a receptor; Adenosine A2a-receptor; Adenosine receptor A2a; adenosine receptor subtype A2a; hA2aR
Gene Aliases: A2AAR; A2aR; AA2AR; ADENO; ADORA2; ADORA2A; Adora2l1; RDC8
UniProt ID: (Human) P29274, (Mouse) Q60613, (Rat) P30543
Entrez Gene ID: (Human) 135, (Mouse) 11540, (Rat) 25369
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support