Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: KIYPEWPVV FFMKGLTDST PMPSNSTYPA FSFLSAIDNV FLFPLDMKRL LEDTPLFSWY NQINASAERN WLHISSDASR LAIIWKYGGI YMDTDVISIR PIPEENFLAA QASRYSSNGI FGFLPHHPFL WECMENFVEH YNSDIWGNQG PELMTRMLRV WCKLEDFQEV SDLRCLNISF LHPQRFYPIS YREWRRYYEV WDTEPSFNVS YALHLWNHMN QEGRAVIRGS NTLVENLYRK HCPRTYRDLI KGPEGSVTGE LGPGNK (76-340 aa encoded by BC119639)
This gene encodes a protein from the glycosyltransferase 32 family. The enzyme catalyzes the transfer of N-acetylglucosamine to core 2 branched O-glycans. It forms a unique glycan, GlcNAcalpha1->4Galbeta->R and is largely associated with the Golgi apparatus membrane.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Alpha-1,4-N-acetylglucosaminyltransferase; alpha4 GnT; Alpha4GnT; GnTalpha4; MGC149493
Gene Aliases: A4GNT; Alpha4gnt; AV080780; Gm798
UniProt ID: (Human) Q9UNA3, (Mouse) Q14BT6
Entrez Gene ID: (Human) 51146, (Rat) 685758, (Mouse) 333424
Molecular Function:
transferase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support