Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Activates acetoacetate to acetoacetyl-CoA. May be involved in utilizing ketone body for the fatty acid-synthesis during adipose tissue development.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: acetoacetate-CoA ligase; Acetoacetyl-CoA synthetase; Acyl-CoA synthetase family member 1; homolog of C. elegans supressor of ras 5 (sur-5); Protein sur-5 homolog
Gene Aliases: AACS; ACSF1; SUR-5
UniProt ID: (Human) Q86V21
Entrez Gene ID: (Human) 65985
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support