Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MKKFFDSRRE QGGSGLGSGS SGGGGSTSGL GSGYIGRVFG IGRQQVTVDE VLAEGGFAIV FLVRTSNGMK CALKRMFVNN EHDLQVCKRE IQIMRDLSGH KNIVGYIDSS INNVSSGDVW EVLILMDFCR GGQVVNLMNQ RLQTGFTENE VLQIFCDTCE AVARLHQCKT PIIHRDLKVE NILLHDRGHY VLCDFGSATN KFQNPQTEGV NAVEDEIKKY TTLSYRAPEM VNLYSGKIIT TKADIWALGC LLYKLCYFTL PFGESQVAIC DGNFTIPDNS RYSQDMHCLI RYMLEPDPDK RPDIYQVSYF SFKLLKKECP IPNVQNSPIP AKLPEPVKAS EAAAKKTQPK ARLTDPIPTT ETSIAPRQRP KAGQTQPNPG ILPIQPALTP RKRATVQPPP QAAGSSNQPG LLASVPQPKP QAPPSQPLPQ TQAKQPQAPP TPQQTPSTQA QGLPAQAQAT PQHQQHTIKL SMKL
AAK1 serine/threonine kinase is a member of the NAK family and plays a regulatory role in endocytosis. AAK1 is a member of the SNF1 subfamily of Ser/Thr protein kinases. The protein interacts with and phosphorylates a subunit of the AP-2 complex, which promotes binding of AP-2 to sorting signals found in membrane-bound receptors and subsequent receptor endocytosis. Its kinase activity is stimulated by clathrin.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Adaptor-associated kinase 1; AP2-associated protein kinase 1; DKFZp686F03202; DKFZp686K16132; FLJ23712; FLJ25931; FLJ31060; FLJ42882; FLJ45252; MGC138170; MGC164568; MGC164570
Gene Aliases: AAK1; KIAA1048
UniProt ID: (Human) Q2M2I8
Entrez Gene ID: (Human) 22848
Molecular Function:
non-receptor serine/threonine protein kinase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support