Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: TINDISVLR VTRRGEQADH FTQTPLDPGS QVLVRVDWER RFDHMQQHSG QHLITAVADH LFKLKTTSWE LGRFRSAIEL DTPSMTAEQV AAIEQSVNEK IRDRLPVNVR ELSLDDPEVE QVSGRGLPDD HAGPIRVVNI EGVDSNMCCG THVSNLSDLQ VIKILGTEKG KKNRTNLIFL SGNRVLKWME RSHGTEKALT ALLKCGAEDH VEAVKKLQNS TKILQKNNLN LLRDLAVHIA HSLRNSPDWG GVVILHRKEG DSEFMNIIAN EIGSEETLLF LTVGDEKGGG LFLLAGPPAS VETLGPRVAE VLEGKGAGKK GRFQGKATKM SRRMEAQALL QDYISTQSAK E (176-525 aa encoded by BC004172)
AARSD1 functions in trans to edit the amino acid moiety from incorrectly charged tRNA(Ala).
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Alanyl-tRNA deacylase alaX; Alanyl-tRNA editing protein Aarsd1; Alanyl-tRNA synthetase domain-containing protein 1; AlaX; MGC2744
Gene Aliases: 1110069E20Rik; 2310044P18Rik; AA589600; AARSD1; AlaX; AlaXp-II
UniProt ID: (Human) Q9BTE6, (Rat) Q5XI97, (Mouse) Q3THG9
Entrez Gene ID: (Human) 80755, (Rat) 619440, (Mouse) 69684
Molecular Function:
RNA metabolism protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support