Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: LALQLEQLLN PRPSEADPEA DPEEATAARV IDRFDEGEDG EGDFLVVGSI RKLASASLLD TDKRYCGKTT SRKAWNEDHW EQTLPGSSDE EISDEEGSGD EDSEGLGLEE YDEDDLGAAE EQECGDHRES KKSRSHSAKT PGFSVQSISD FEKFTKGMDD LGSSEEEEDE ESGMEEGDDA EDSQGESEED RAGDRNSEDD GVVMTF
The protein encoded by this gene was identified on the basis of its interaction with MAP3K12/DLK, a protein kinase known to be involved in the induction of cell apoptosis. This gene product contains a leucine zipper, which is a characteristic motif of transcription factors, and was shown to exhibit strong transactivation activity when fused to Gal4 DNA binding domain. Overexpression of this gene interfered with MAP3K12 induced apoptosis.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Apoptosis-antagonizing transcription factor; Protein AATF; Rb-binding protein Che-1; Traube protein
Gene Aliases: 4933415H02Rik; 5830465M17Rik; AATF; BFR2; CHE-1; CHE1; DED; HSPC277; Trb
UniProt ID: (Human) Q9NY61, (Mouse) Q9JKX4, (Rat) Q9QYW0
Entrez Gene ID: (Human) 26574, (Mouse) 56321, (Rat) 114512
Molecular Function:
chromatin/chromatin-binding, or -regulatory protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support