Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Peptide Sequence: MNKMALASFM KGRTVIGTPD EETMDIELPK KYHEMVGVIF SDTFSYRLKF NWGYRIPVIK EHSEYTEHCW AMHGEIFCYL
ABCA10 (ATP Binding Cassette Subfamily A Member 10) is a Protein Coding gene. Diseases associated with ABCA10 include Donnai-Barrow Syndrome. Among its related pathways are Regulation of activated PAK-2p34 by proteasome mediated degradation and Transport of glucose and other sugars, bile salts and organic acids, metal ions and amine compounds. Gene Ontology (GO) annotations related to this gene include ATPase activity and ATPase activity, coupled to transmembrane movement of substances. An important paralog of this gene is ABCA9.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: ATP-binding cassette A10; ATP-binding cassette sub-family A member 10; ATP-binding cassette, sub-family A (ABC1), member 10; FKHL6; FREAC2
Gene Aliases: ABCA10; EST698739
UniProt ID: (Human) Q8WWZ4
Entrez Gene ID: (Human) 10349
Molecular Function:
ATP-binding cassette (ABC) transporter
primary active transporter
transmembrane signal receptor
transporter
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support