Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 3
Positive test controls include: HeLa, Rat brain. The target is usually found in the following locations: Endosome membrane, Lysosome membrane, Multi-pass membrane protein.
Immunogen sequence: SHLDRSTVSS FSLDSVARNP QELWRFLTQN LSLPNSTAQA LLAARVDPPE VYHLLFGPSS ALDSQSGLHK GQEPWSRLGG NPLFRMEELL LAPALLEQLT CTPGSGELGR ILTVPESQKG ALQGYRDAVC SGQAAARARR FSGLSAELRN QLDVAKVSQQ LGLDAPNGSD SSPQAPPPRR LQALLGDLLD AQKVLQDVDV LSALALLLPQ G
Probable transporter, its natural substrate has not been found yet. May have a role in macrophage lipid metabolism and neural development.
⚠WARNING: This product can expose you to chemicals including mercury, which is known to the State of California to cause birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: abca2 {ECO:0000312|RGD:620238}; ATP-binding cassette 2; ATP-binding cassette sub-family A member 2; ATP-binding cassette transporter 2; ATP-binding cassette, sub-family A (ABC1), member 2; ATP-binding cassette, sub-family A, member 2
Gene Aliases: ABC2; ABCA2; AI413825; D2H0S1474E; KIAA1062; mKIAA1062
UniProt ID: (Human) Q9BZC7, (Rat) Q9ESR9
Entrez Gene ID: (Human) 20, (Mouse) 11305, (Rat) 79248
Molecular Function:
ATP-binding cassette (ABC) transporter
primary active transporter
transporter
non-motor actin binding protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support