Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Reconstitute in 100 µL of sterile water. Centrifuge to remove any insoluble material.
Specificity of this antibody: ABCA3.
Plays an important role in the formation of pulmonary surfactant, probably by transporting lipids such as cholesterol.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: ABC transporter 3; ABC-C transporter; ATP-binding cassette 3; ATP-binding cassette sub-family A member 3; ATP-binding cassette transporter 3; ATP-binding cassette, sub-family A (ABC1), member 3; Phospholipid-transporting ATPase ABCA3; Xenobiotic-transporting ATPase ABCA3
Gene Aliases: ABC-C; ABC3; ABCA3; EST111653; LBM180; SMDP3
UniProt ID: (Human) Q99758
Entrez Gene ID: (Human) 21
Molecular Function:
ATP-binding cassette (ABC) transporter
primary active transporter
transporter
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support