Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: SLLTMDLGR VDTFNESRFS VVYTPVTNTT QQIMNKVAST PFLAGKEVLG LPDEESIKEF TANYPEEIVR VTFTNTYSYH LKFLLGHGMP AKKEHKDHTA HCYETNEDVY CEVSVFWKEG FVALQAAI (58-184 aa encoded by bc130280 )
ATP-binding cassette (ABC) transporters are an evolutionarily conserved family of proteins that catalyze the transport of molecules across extracellular and intracellular membranes by harnessing the energy of ATP hydrolysis. ABCA8 (ATP-binding cassette, sub-family A (ABC1), member 8) is a 1, 581 amino acid multi-pass membrane protein that contains two ABC transporter domains and belongs to the ABC transporter family. Expressed in a variety of tissues with particularly high expression in liver, heart and skeletal muscle, ABCA8 functions as an ATP-dependent lipophilic drug transporter that exists as multiple alternatively spliced isoforms.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: ABC-type organic anion transporter ABCA8; ABC-type organic anion transporter ABCA8A; ABC-type organic anion transporter ABCA8B; ATP-binding cassette sub-family A member 8; ATP-binding cassette sub-family A member 8-A; ATP-binding cassette sub-family A member 8-B; ATP-binding cassette sub-family A member 8A; ATP-binding cassette sub-family A member 8B; ATP-binding cassette, sub-family A (ABC1), member 8
Gene Aliases: ABCA8; Abca8a; Abca8b; KIAA0822
UniProt ID: (Human) O94911, (Mouse) Q8K442, (Mouse) Q8K440
Entrez Gene ID: (Human) 10351, (Mouse) 217258, (Mouse) 27404
Molecular Function:
ATP-binding cassette (ABC) transporter
primary active transporter
transporter
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support