Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: FVPIAVKGLT NSKSAVMRFK KFFLQESPVF YVQTLQDPSK ALVFEEATLS WQQTCPGIVN GALELERNGH ASEGMTRPRD ALGPEEEGNS LGPELHKINL
ABCC11 is a member of the superfamily of ATP-binding cassette (ABC) transporters. ABC proteins transport various molecules across extra- and intra-cellular membranes. ABC genes are divided into seven distinct subfamilies (ABC1, MDR/TAP, MRP, ALD, OABP, GCN20, White). This ABC full transporter is a member of the MRP subfamily which is involved in multi-drug resistance. The product of this gene participates in physiological processes involving bile acids, conjugated steroids, and cyclic nucleotides. In addition, a SNP in this gene is responsible for determination of human earwax type. This gene and family member ABCC12 are determined to be derived by duplication and are both localized to chromosome 16q12.1. Multiple alternatively spliced transcript variants have been described for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: ATP-binding cassette protein C11; ATP-binding cassette sub-family C member 11; ATP-binding cassette transporter MRP8; ATP-binding cassette transporter sub-family C member 11; ATP-binding cassette, sub-family C (CFTR/MRP), member 11; multi-resistance protein 8; Multidrug resistance-associated protein 8
Gene Aliases: ABCC11; EWWD; MRP8; WW
UniProt ID: (Human) Q96J66
Entrez Gene ID: (Human) 85320
Molecular Function:
ATP-binding cassette (ABC) transporter
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support