Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 2
Antibody detects endogenous levels of total ABCC9.
Sulfonylurea receptors (SUR) are membrane proteins which are the molecular targets of the sulfonylurea class of anti-diabetic drugs whose mechanism of action is to promote insulin release from pancreatic beta cells. More specifically, SUR proteins are subunits of the inward-rectifier potassium ion channels Kir6.x (6.1 and 6.2). The association of four Kir6.x and four SUR subunits form an ion conducting channel commonly referred to as the KATP channel. The primary function of the sulfonylurea receptor is to sense intracellular levels of the nucleotides ATP and ADP and in response facilitate the open or closing its associated Kir6.x potassium channel. Hence the KATP channel monitors the energy balance within the cell.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: ATP-binding cassette sub-family C member 9; ATP-binding cassette transporter sub-family C member 9; ATP-binding cassette, sub-family C (CFTR/MRP), member 9; FLJ36852; Sulfonylurea receptor 2
Gene Aliases: ABC37; ABCC9; ATFB12; CANTU; CMD1O; SUR2
UniProt ID: (Human) O60706, (Rat) Q63563
Entrez Gene ID: (Human) 10060, (Rat) 25560
Molecular Function:
ATP-binding cassette (ABC) transporter
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support