Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: SESDAEAVK KAALEKKEEE LVSERTEAFT IARNLLTAAA DAIERIMSSY KEVTELAGYT ARVHEMFQVF EDVQRCHFKR PRELEDAQAG SGTIGRSGVR VEGPLKIRGQ VVDVEQGIIC ENIPIVTPSG EVVVASLNIR VEEGMHLLIT GPNGCGKSSL FRILGGLWPT YGGVLYKPPP QRMFYIPQRP YMSVGSLRDQ VIYPDSVEDM QRKGYSEQDL EAILDVVHLH HILQREGGWE AMCDWKDVLS GGEKQRIGMA RMFYHRPKYA LLDECTSAVS IDVEGKIFQA AKDAGIALLS ITHRPSLWKY HTHLLQFDGE GGWKFEKLDS AARLSLTEEK QRLEQQLAGI PKMQRRLQEL CQILGEAVAP AHVPAPSPQG PGGLQGAST (358-745 aa encoded by BC025358)
This gene is a member of the UBA domain family, whose members include proteins having connections to ubiquitin and the ubiquitination pathway. The ubiquitin associated domain is thought to be a non-covalent ubiquitin binding domain consisting of a compact three helix bundle. This particular protein originates from a gene locus in a refined region on chromosome 9 undergoing loss of heterozygosity in nasopharyngeal carcinoma (NPC). Taking into account its cytogenetic location, this UBA domain family member is being studies as a putative target for mutation in nasopharyngeal carcinomas. Multiple alternatively spliced transcript variants encoding different isoforms have been found for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Adrenoleukodystrophy protein; ALDP; ATP-binding cassette sub-family D member 1; ATP-binding cassette, sub-family D (ALD), member 1
Gene Aliases: ABC42; ABCD1; ALD; ALDP; AMN
UniProt ID: (Human) P33897
Entrez Gene ID: (Human) 215
Molecular Function:
ATP-binding cassette (ABC) transporter
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support