Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Reconstitute in 100 µL of sterile water. Centrifuge to remove any insoluble material.
Specificity of this antibody: ABCE1.
The protein encoded by this gene is a member of the superfamily of ATP-binding cassette (ABC) transporters. ABC proteins transport various molecules across extra- and intra-cellular membranes. ABC genes are divided into seven distinct subfamilies (ABC1, MDR/TAP, MRP, ALD, OABP, GCN20, White). This protein is a member of the OABP subfamily. Alternatively referred to as the RNase L inhibitor, this protein functions to block the activity of ribonuclease L. Activation of ribonuclease L leads to inhibition of protein synthesis in the 2-5A/RNase L system, the central pathway for viral interferon action. Two transcript variants encoding the same protein have been found for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 2'-5'-oligoadenylate-binding protein; ABC 38; ABCE 1; ATP-binding cassette sub-family E member 1; ATP-binding cassette, sub-family E (OABP), member 1; ATP-binding cassette, subfamily E, member 1; HP68; HuHP68; Ribonuclease 4 inhibitor; ribonuclease L (2', 5'-oligoisoadenylate synthetase-dependent) inhibitor; ribonuclease L (2',5'-oligoisoadenylate synthetase-dependent) inhibitor; RNase 4 inhibitor; RNase L inhibitor; RNase L inhibitor 1; RNS4I; RNS4l (Eye)
Gene Aliases: ABC38; ABCE1; C79080; OABP; OK/SW-cl.40; RLI; RLI1; RNASEL1; RNASELI; RNS41; RNS4I
UniProt ID: (Human) P61221, (Mouse) P61222
Entrez Gene ID: (Human) 6059, (Mouse) 24015
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support