Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Sequence of this protein is as follows: MAAEEEEVDS ADTGERSGWL TGWLPTWCPT SISHLKEAEE KMLKCVPCTY KKEPVRISNG NKIWTLKFSH NISNKTPLVL LHGFGGGLGL WALNFGDLCT NRPVYAFDLL GFGRSSRPRF DSDAEEVENQ FVESIEEWRC ALGLDKMILL GHNLGGFLAA AYSLKYPSRV NHLILVEPWG FPERPDLADQ DRPIPVWIRA LGAALTPFNP LAGLRIAGPF GLSLVQRLRP DFKRKYSSMF EDDTVTEYIY HCNVQTPSGE TAFKNMTIPY GWAKRPMLQR IGKMHPDIPV SVIFGARSCI DGNSGTSIQS LRPHSYVKTI AILGAGHYVY ADQPEEFNQK VKEICDTVD
The protein encoded by this gene belongs to a large family of proteins defined by an alpha/beta hydrolase fold, and contains three sequence motifs that correspond to a catalytic triad found in the esterase/lipase/thioesterase subfamily. It differs from other members of this subfamily in that its putative catalytic triad contains an asparagine instead of the serine residue. Mutations in this gene have been associated with Chanarin-Dorfman syndrome, a triglyceride storage disease with impaired long-chain fatty acid oxidation.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 1-acylglycerol-3-phosphate O-acyltransferase ABHD5; Abhydrolase domain-containing protein 5; aCGI58; CGI 58; CGI58 protein; Lipid droplet-binding protein CGI-58; MGC8731; Protein CGI-58; truncated abhydrolase domain-containing protein 5
Gene Aliases: ABHD5; CDS; CGI-58; CGI58; IECN2; NCIE2
UniProt ID: (Human) B2R9K0, (Rat) Q6QA69
Entrez Gene ID: (Human) 51099, (Rat) 316122
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support