Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Antibody detects endogenous levels of total ABHD6.
ABHD6 has 2-arachidonoylglycerol hydrolase activity. ABHD6 may be a regulator of endocannabinoid signaling pathways.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 2-arachidonoylglycerol hydrolase; Abhydrolase domain-containing protein 6; lipase protein; Monoacylglycerol lipase ABHD6
Gene Aliases: 0610041D24Rik; AA673485; ABHD6; AV065425
UniProt ID: (Human) Q9BV23, (Rat) Q5XI64, (Mouse) Q8R2Y0
Entrez Gene ID: (Human) 57406, (Rat) 305795, (Mouse) 66082
Molecular Function:
lipase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support