Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
The recommended shelf life for this product is 1 year from date of receipt.
Application Note: For IHC, epitope retrieval with citrate buffer pH 6.0 is recommended for FFPE tissue sections.
Based on 100% sequence identity, this antibody is predicted to react with Mouse and Rat
This gene encodes a member of the DEAD box protein family. DEAD box proteins, characterized by the conserved motif Asp-Glu-Ala-Asp (DEAD), are putative RNA helicases. They are implicated in a number of cellular processes involving alteration of RNA secondary structure such as translation initiation, nuclear and mitochondrial splicing, and ribosome and spliceosome assembly. Based on their distribution patterns, some members of this family are believed to be involved in embryogenesis, spermatogenesis, and cellular growth and division. The nucleolar protein encoded by this gene interacts in a hormone-dependent manner with nuclear receptors, and represses their transcriptional activity. Alternative splice variants that encode different isoforms have been found for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Abelson interactor 1; Abi-1; Abl interactor 1; Abl-binding protein 4; abl-interactor 1; Abl-interactor protein 1 long; AblBP4; e3B1; EL2; Eps8 SH3 domain-binding protein; Eps8-binding protein; HS3; interactor protein AblBP4; nap1 binding protein; Nap1-binding protein; Nap1BP; Spectrin SH3 domain-binding protein 1; SPH3; SPTA
Gene Aliases: ABI-1; ABI1; ABLBP4; E3B1; NAP1BP; SSH3BP; SSH3BP1
UniProt ID: (Human) Q8IZP0
Entrez Gene ID: (Human) 10006
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support