Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: DIERPDLIT YEPFYTSGYD DKQERQSLGE SPRTLSPTPS AEGYQDVRDR MIHRSTSQGS INSPVYSRHS YTPTTSRSPQ HFHRPDQGIN IYRKPPIYKQ HAALAAQSKS SEDIIKFSKF PAAQAPDPSE TPKIETDHWP GPPSFAVVGP DMKRRSSGRE EDDEELLRRR QLQEEQLMKL NSGLGQLILK EEMEKESRER SSLLASRYDS PINSASHIPS SKTASLPGYG RNGLHRPVST DFAQYNSYGD VSGGVRDYQT LPDGHMPAMR MDRGVSMPNM LEPKIFPYEM LMVTNRGRNK ILREVDRTRL ERHLAPEVFR EIFGMSIQEF DRLPLWRRND MKKKAKLF (55-401 aa encoded by BC002448 )
This gene encodes a cytoskeletal LIM protein that binds to actin filaments via a domain that is homologous to erythrocyte dematin. LIM domains, found in over 60 proteins, play key roles in the regulation of developmental pathways. LIM domains also function as protein-binding interfaces, mediating specific protein-protein interactions. The protein encoded by this gene could mediate such interactions between actin filaments and cytoplasmic targets. Alternatively spliced transcript variants encoding different isoforms have been identified.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: abLIM-1; Actin-binding double zinc finger protein; actin-binding double-zinc-finger protein; Actin-binding LIM protein 1; Actin-binding LIM protein family member 1; LIMAB1; Limatin
Gene Aliases: 2210411C18Rik; 2610209L21Rik; 4833406P10Rik; 9330196J19Rik; ABLIM; abLIM-1; ABLIM1; AV079770; AW060987; AW215784; KIAA0059; LIMAB1; LIMATIN; mKIAA0059; RGD1565768
UniProt ID: (Human) O14639, (Mouse) Q8K4G5
Entrez Gene ID: (Human) 3983, (Rat) 307989, (Mouse) 226251
Molecular Function:
actin or actin-binding cytoskeletal protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support