Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: ESDASSQGS GSEGPVRLSR TRTKALQEAM YYSAEHGYVD ITMELRALGV PWKLHIWIES LRTSFSQSRY SVVQSLLRDF SSIREEEYNE ELVTEGLQLM FDILKTSKND SVIQQLATIF THCYGSSPIP SIPEIRKTLP ARLDPHFLNN KEMSDVTFLV EGKLFYAHKV LLVTASNRFK TLMTNKSEQD GDSSKTIEIS DMKYHIFQMM MQYLYYGGTE SMEIPTTDIL ELLSAASLFQ LDALQRHCEI LCSQTLSMES AVNTYKYAKI HNAPELALFC EGFFLKHMKA LLEQDAFRQL IYGRSSKVQG LDPLQDLQNT LAERVHSVYI TSRV (507-839 aa encoded by B C093968)
May be involved in the initiation of hepatocyte growth. Target protein displays a transient increase in mRNA expression prior to DNA synthesis in regenerating liver cells.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: ankyrin repeat and BTB (POZ) domain containing 2; Ankyrin repeat and BTB/POZ domain-containing protein 2
Gene Aliases: ABTB2; ABTB2A; BTBD22
UniProt ID: (Human) Q8N961
Entrez Gene ID: (Human) 25841
Molecular Function:
scaffold/adaptor protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support