Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
This Antibody was verified by Knockdown to ensure that the antibody binds to the antigen stated.
FIGURE: 1 / 1
Immunogen sequence: VLKDVNLRPE QLGDICVGNV LQPGAGAIMA RIAQFLSDIP ETVPLSTVNR QCSSGLQAVA SIAGGIRNGS YDIGMACGVE SMSLADRGNP GNITSRLMEK EKARDCLIPM GITSENVAER FGISREKQ
This gene encodes an enzyme operative in the beta-oxidation system of the peroxisomes. Deficiency of this enzyme leads to pseudo-Zellweger syndrome. Alternative splicing results in multiple transcript variants.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 3-ketoacyl-CoA thiolase, peroxisomal; Acetyl-CoA acyltransferase; Acetyl-CoA C-myristoyltransferase; acetyl-Coenzyme A acyltransferase 1; Beta-ketothiolase; Peroxisomal 3-oxoacyl-CoA thiolase; peroxisomal 3-oxoacyl-Coenzyme A thiolase; testicular tissue protein Li 197
Gene Aliases: ACAA; ACAA1; PTHIO; THIO
UniProt ID: (Human) P09110
Entrez Gene ID: (Human) 30
Molecular Function:
acetyltransferase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support