Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: EERLETPSA KKLTDIGIRR IFSPEHDIFR KSVRKFFQEE VIPHHSEWEK AGEVSREVWE KAGKQGLLGV NIAEHLGGIG GDLYSAAIVW EEQAYSNCSG PGFSIHSGIV MSYITNHGSE EQIKHFIPQM TAGKCIGAIA MTEPGAGSDL QGIKTNAKKD GSDWILNGSK VFISNGSLSD VVIVVAVTNH EAPSPAHGIS LFLVENGMKG FIKGRKLHKM GLKAQDTAEL FFEDIRLPAS ALLGEENKGF YYIMKELPQE RLLIADVAIS ASEFMFEETR NYVKQRKAFG KTVAHLQTVQ HKLAELKTHI CVTRAFVDNC LQLHEAKRLD SATACMAKYW ASELQNSVAY DCVQLHGGWG YMWEYPIAKA YVDARVQPIY GGTNEIMKEL IAREIVFDK (33-430 aa encoded by BC039063)
The protein encoded by this gene belongs to the acyl-CoA dehydrogenase family, which is a family of mitochondrial flavoenzymes involved in fatty acid and branched chain amino-acid metabolism. This protein is one of the four enzymes that catalyze the initial step of mitochondrial beta-oxidation of straight-chain fatty acid. Defects in this gene are the cause of long-chain acyl-CoA dehydrogenase (LCAD) deficiency, leading to nonketotic hypoglycemia.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: acetyl-Coenzyme A dehydrogenase, long-chain; Acyl Coenzyme A dehydrogenase, long chain; acyl-Coenzyme A dehydrogenase, long chain; acyl-Coenzyme A dehydrogenase, long-chain; LCAD; LCAD long chain acyl-CoA dehydrogenase; long-chain acyl-CoA dehydrogenase; Long-chain specific acyl-CoA dehydrogenase, mitochondrial
Gene Aliases: AA960361; ACAD4; ACADL; ACOADA; AU018452; C79855; LCAD
UniProt ID: (Human) P28330, (Mouse) P51174, (Rat) P15650
Entrez Gene ID: (Human) 33, (Mouse) 11363, (Rat) 25287
Molecular Function:
dehydrogenase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support