Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 4
Positive Samples: SH-SY5Y, Mouse liver, Mouse kidney, Mouse heart, Rat spinal cord
Immunogen sequence: GGEERLETPS AKKLTDIGIR RIFSPEHDIF RKSVRKFFQE EVIPHHSEWE KAGEVSREVW EKAGKQGLLG VNIAEHLGGI GGDLYSAAIV WEEQAYSNCS GPGFSIHSGI VMSYITNHGS EEQIKHFIPQ MTAGKCIGAI AMTEPGAGSD LQGIKTNAKK DGSDWILNGS KVFISNGSLS
The protein encoded by this gene belongs to the acyl-CoA dehydrogenase family, which is a family of mitochondrial flavoenzymes involved in fatty acid and branched chain amino-acid metabolism. This protein is one of the four enzymes that catalyze the initial step of mitochondrial beta-oxidation of straight-chain fatty acid. Defects in this gene are the cause of long-chain acyl-CoA dehydrogenase (LCAD) deficiency, leading to nonketotic hypoglycemia.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: acetyl-Coenzyme A dehydrogenase, long-chain; Acyl Coenzyme A dehydrogenase, long chain; acyl-Coenzyme A dehydrogenase, long chain; acyl-Coenzyme A dehydrogenase, long-chain; LCAD; LCAD long chain acyl-CoA dehydrogenase; long-chain acyl-CoA dehydrogenase; Long-chain specific acyl-CoA dehydrogenase, mitochondrial
Gene Aliases: AA960361; ACAD4; ACADL; ACOADA; AU018452; C79855; LCAD
UniProt ID: (Human) P28330, (Mouse) P51174, (Rat) P15650
Entrez Gene ID: (Human) 33, (Mouse) 11363, (Rat) 25287
Molecular Function:
dehydrogenase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support