Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Cell lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MAAGFGRCCR VLRSISRFHW RSQHTKANRQ REPGLGFSFE FTEQQKEFQA TARKFAREEI IPVAAEYDKT GEYPVPLIRR AWELGLMNTH IPENCGGLGL GTFDACLISE ELAYGCTGVQ TAIEGNSLGQ MPIIIAGNDQ QKKKYLGRMT EEPLMCAYCV TEPGAGSDVA GIKTKAEKKG DEYIINGQKM WITNGGKANW YFLLARSDPD PKAPANKAFT GFIVEADTPG IQIGRKELNM GQRCSDTRGI VFEDVKVPKE NVLIGDGAGF KVAMGAFDKT RPVVAAGAVG LAQRALDEAT KYALERKTFG KLLVEHQAIS FMLAEMAMKV ELARMSYQRA AWEVDSGRRN TYYASIAKAF AGDIANQLAT DAVQILGGNG FNTEYPVEKL MRDAKIYQIY EGTSQIQRLI VAREHIDKYK N
This gene encodes the medium-chain specific (C4 to C12 straight chain) acyl-Coenzyme A dehydrogenase. The homotetramer enzyme catalyzes the initial step of the mitochondrial fatty acid beta-oxidation pathway. Defects in this gene cause medium-chain acyl-CoA dehydrogenase deficiency, a disease characterized by hepatic dysfunction, fasting hypoglycemia, and encephalopathy, which can result in infantile death. Alternatively spliced transcript variants encoding different isoforms have been found for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: acyl-CoA dehydrogenase C-4 to C-12 straight chain; acyl-Coenzyme A dehydrogenase, C-4 to C-12 straight chain; FLJ18227; FLJ93013; FLJ99884; MCAD; MCADH; Medium chain acyl-CoA dehydrogenase; Medium-chain specific acyl-CoA dehydrogenase, mitochondrial; RP4-682C21.1; testicular tissue protein Li 7
Gene Aliases: ACAD1; ACADM; MCAD; MCADH
UniProt ID: (Human) P11310
Entrez Gene ID: (Human) 34
Molecular Function:
dehydrogenase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support