Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: WYFLLARSDP DPKAPANKAF TGFIVEADTP GIQIGRKELN MGQRCSDTRG IVFEDVKVPK ENVLIGDGAG FKVAMGAFDK TRPVVAAGAV GLAQRALDEA T
This gene encodes the medium-chain specific (C4 to C12 straight chain) acyl-Coenzyme A dehydrogenase. The homotetramer enzyme catalyzes the initial step of the mitochondrial fatty acid beta-oxidation pathway. Defects in this gene cause medium-chain acyl-CoA dehydrogenase deficiency, a disease characterized by hepatic dysfunction, fasting hypoglycemia, and encephalopathy, which can result in infantile death. Alternatively spliced transcript variants encoding different isoforms have been found for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: acyl-CoA dehydrogenase C-4 to C-12 straight chain; Acyl-Coenzyme A dehydrogenase C-4 to C-12 straight-chain; acyl-Coenzyme A dehydrogenase, C-4 to C-12 straight chain; Acyl-Coenzyme A dehydrogenase, C-4 to C-12 straight-chain; acyl-Coenzyme A dehydrogenase, medium chain; FLJ18227; FLJ93013; FLJ99884; MCAD; MCADH; Medium chain acyl-CoA dehydrogenase; medium-chain acyl-CoA dehydrogenase; Medium-chain specific acyl-CoA dehydrogenase, mitochondrial; RP4-682C21.1; testicular tissue protein Li 7
Gene Aliases: ACAD1; ACADM; AU018656; MCAD; MCADH
UniProt ID: (Human) P11310, (Mouse) P45952, (Rat) P08503
Entrez Gene ID: (Human) 34, (Mouse) 11364, (Rat) 24158
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support