Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MTTLLWVFVT LRVITAAVTV ETSDHDNSLS VSIPQPSPLR VLLGTSLTIP CYFIDPMHPV TTAPSTAPLA PRIKWSRVSK EKEVVLLVAT EGRVRVNSAY QDKVSLPNYP AIPSDATLEV QSLRSNDSGV YRCEVMHGIE DSEATLEVVV KGIVFHYRAI STRYTLDFDR AQRACLQNSA IIATPEQLQA AYEDGFHQCD AGWLADQTVR YPIHTPREGC YGDKDEFPGV RTYGIRDTNE TYDVYCFAEE MEGEVFYATS PEKFTFQEAA NECRRLGARL ATTGQLYLAW QAGMDMCSAG WLADRSVRYP ISKARPNCGG NLLGVRTVYV HANQTGYPDP SSRYDAICYT GEDFVDIPEN FFGVGGEEDI TVQTVTWPDM ELPLPRNITE GEARGSVILT VKPIFEVSPS PLEPEEPFTF APEIGATAFA EVENETGEAT RPWGFPTPGL GPATAFTSED LVVQVTAVPG QPHLPGGVVF HYRPGPTRYS LTFEEAQQAC LRTGAVIASP EQLQAAYEAG YEQCDAGWLR DQTVRYPIVS PRTPCVGDKD SSPGVRTYGV RPSTETYDVY CFVDRLEGEV FFATRLEQFT FQEALEFCES HNATLATTGQ LYAAWSRGLD KCYAGWLADG SLRYPIVTPR PACGGDKPGV RTVYLYPNQT GLPDPLSRHH AFCFRGMQPH FGSNSPFCLE RTPLGSPDPA TTQYPIHQSK KMSVPLGQSQ L
Aggrecan also known as cartilage-specific proteoglycan core protein (CSPCP) or chondroitin sulfate proteoglycan 1 is a protein that is encoded by the ACAN gene. The encoded protein is an integral part of the extracellular matrix in cartilaginous tissue and it withstands compression in cartilage. The human form of the protein is 2316 amino acids long (approximately 250 kDa) and can be expressed in multiple isoforms due to alternative splicing. Aggrecan exhibits a bottlebrush structure, in which chondroitin sulfate and keratan sulfate chains are attached to an extended protein core. The synthesis and degradation of aggrecan are being investigated for their roles in cartilage deterioration during joint injury, disease, and aging. It contains three interglobular domains (IGDs), G1, G2, and G3 that are involved in aggregation, hyaluronan binding, cell adhesion, and chondrocyte apoptosis. Aggrecan fragments from articular cartilage are released into the synovial fluid at all stages of human osteoarthritis as a result of cleavage by aggrecanase and MMP activity. Antibodies BC-3 and BC-4 can be used to detect these neo epitopes of Aggrecanase (ARGSVXXX) and MMP (XXXVDIPEN), respectively.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Aggrecan core protein; Cartilage-specific proteoglycan core protein; Chondroitin sulfate proteoglycan 1; Chondroitin sulfate proteoglycan core protein 1; CSPCP; large aggregating proteoglycan
Gene Aliases: ACAN; AGC1; AGCAN; CSPG1; CSPGCP; MSK16; SEDK
UniProt ID: (Human) P16112
Entrez Gene ID: (Human) 176
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support