Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: CEGAGSRKP TASSSRQDKE AWIKDKYVEK KFLRKAPMAP ALEAPRRWRV QKCLRPHSSP RAPTARRKVR LEPVLPCVAA LSSVGTLDRK FRRDSLFCPD ELDSLFSYFD AGAAGAGPRS LSSDSGLGGS SDGSSDVLAF GSGSVVDSVT EEEGAESEES SGEADGDTEA EAWGLADVRE LHPGLLAHRA ARARDLPALA AALAHGAEVN WADAEDEGKT PLVQAVLGGS LIVCEFLLQN GADVNQRDSR GRAPLHHATL LGRTGQVCLF LKRGADQHAL DQEQRDPLAI AVQAANADIV TLLRLARMAE EMREAEAAPG PPGALAGSPT ELQFRRCIQE FISLHLEES (217-564 aa encoded by BC047001)
ACAP3 is GTPase-activating protein for the ADP ribosylation factor family.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Arf-GAP with coiled-coil, ANK repeat and PH domain-containing protein 3; centaurin; centaurin, beta 5; Centaurin-beta-5; Cnt-b5
Gene Aliases: ACAP3; CENTB5; KIAA1716; Kiaa1716-hp; mKIAA1716
UniProt ID: (Human) Q96P50
Entrez Gene ID: (Human) 116983, (Mouse) 140500, (Rat) 313772
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support