Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Peptide sequence: MGTEKESPEP DCQKQFQAAV SVIQNLPKNG SYRPSYEEML RFYSYYKQAT
Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 79%; Horse: 100%; Human: 100%; Mouse: 100%; Pig: 93%; Rabbit: 100%; Rat: 100%; Zebrafish: 85%
ACBD4 is a member of the acyl-coenzyme A binding domain containing protein family. All family members contain the conserved acyl-Coenzyme A binding domain, which binds acyl-CoA thiol esters. They are thought to play roles in acyl-CoA dependent lipid metabolism.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Acyl-CoA-binding domain-containing protein 4; acyl-Coenzyme A binding domain containing 4; FLJ13322; FLJ90623
Gene Aliases: ACBD4; HMFT0700
UniProt ID: (Human) Q8NC06
Entrez Gene ID: (Human) 79777
Molecular Function:
transfer/carrier protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support