Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
Immunogen sequence: MFSSEVQFGH AGACANQASE TAVAKNQALK EAGVFVPRSF DELGEIIQSV YEDLVANGVI VPAQEVPPPT VPMDYSWARE LGLIRKPASF MTSICDERGQ ELIYAGMPIT EVFKEEMGIG GVLGLLWFQK RLPKYSCQFI EMCLMVTADH GPAVSGAHNT IICARAGKDL VSSLTSGLLT IGDRFGGALD AAAKMFSKAF DSGIIPMEFV NKMKKEGKLI MGIGHRVKSI NNPDMRVQIL KDYVRQHFPA TPLLDYALEV EKITTSKKPN LILNVDGLIG VAFVDMLRNC GSFTREEADE YIDIGALNGI FVLGRSMGFI GHYLDQKRLK QGLYRHPWDD ISYVLPEHMS M
ATP citrate lyase is the primary enzyme responsible for the synthesis of cytosolic acetyl-CoA in many tissues. The enzyme is a tetramer (relative molecular weight approximately 440,000) of apparently identical subunits. It catalyzes the formation of acetyl-CoA and oxaloacetate from citrate and CoA with a concomitant hydrolysis of ATP to ADP and phosphate. The product, acetyl-CoA, serves several important biosynthetic pathways, including lipogenesis and cholesterogenesis. In nervous tissue, ATP citrate-lyase may be involved in the biosynthesis of acetylcholine. Two transcript variants encoding distinct isoforms have been identified for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: ACL; ATP-citrate (pro-S-)-lyase; ATP-citrate synthase; Citrate cleavage enzyme
Gene Aliases: A730098H14Rik; ACL; ACLY; ATPCL; AW538652; CLATP
UniProt ID: (Human) P53396, (Mouse) Q91V92, (Rat) P16638
Entrez Gene ID: (Human) 47, (Mouse) 104112, (Rat) 24159
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support