Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 3
Positive test controls include: 293T, LO2, U-87MG, Mouse kidney. The target is usually found in the following locations: Mitochondrion.
Immunogen sequence: IVDMFGTGGG LLEYRASLLA GKGFAVMALA YYNYEDLPKT METLHLEYFE EAMNYLLSHP EVKGPGVGLL GISKGGELCL SMASFLKGIT AAVVINGSVA NVGGTLHYKG ETLPPVGVNR NRIKVTKDGY ADIVDVLNSP LEGPDQKSFI PVERAESTFL FLVGQDDHNW KSEFYANEAC KRLQAHGRRK PQIICYPETG HYIEPPYFPL CRASLHALVG SPIIWGGEPR AHAMAQVDAW KQLQTFFHKH LGGHEGTIPS KV
Acyl-CoA thioesterases (ACOTs) are a group of enzymes that catalyze the hydrolysis of acyl-CoA to form coenzyme A (CoA) and a free fatty acid. Through their catalytic activity, ACOTs are able to regulate the level of fatty acids and acyl-CoAs within the cell. ACOT1 (acyl-CoA thioesterase 1, also known as CTE1) and ACOT2 (acyl-CoA thioesterase 2, also known as PTE2) are members of the ACOT family and exhibit different cellular localization, with ACOT1 existing as a monomer in the cytoplasm and ACOT2 localized primarily to mitochondria. Characteristic of most ACOT proteins, ACOT1 and ACOT2 catalyze the conversion of Palmitoyl-CoA and water to free CoA and palmitate, a reaction that is important for the regulation of intercellular fatty acid levels. ACOT2 is expressed as multiple alternatively spliced isoforms and, like ACOT1, is encoded by a gene which maps to human chromosome 14.
⚠WARNING: This product can expose you to chemicals including mercury, which is known to the State of California to cause birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Acyl coenzyme A thioester hydrolase; Acyl-CoA thioesterase 2; Acyl-coenzyme A thioester hydrolase 2a; Acyl-coenzyme A thioesterase 2, mitochondrial; ARTISt/p43; CTE Ia; CTE-Ia; Long-chain acyl-CoA thioesterase 2; mitochondrial acyl-CoA thioesterase 1; MTE-I; peroxisomal long-chain acyl-coA thioesterase 2; Very-long-chain acyl-CoA thioesterase; ZAP128
Gene Aliases: AA571646; ACOT2; CTE-IA; CTE1A; MTE-I; MTE1; PTE2; PTE2A; ZAP128
UniProt ID: (Human) P49753, (Mouse) Q9QYR9
Entrez Gene ID: (Human) 10965, (Mouse) 171210, (Rat) 192272
Molecular Function:
esterase
hydrolase
metabolite interconversion enzyme
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support