Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: PDTCALLQP PAASAAAAPS MSGPDVETPS AIQICRIMRP DDANVAGNVH GGTILKMIEE AGAIISTRHC NSQNGERCVA ALARVERTDF LSPMCIGEVA HVSAEITYTS KHSVEVQVNV MSENILTGAK KLTNKATLWY VPLSLKNVDK VLEVPPVVYS RQEQEEEGRK RYEAQKLERM ETKWRNGDIV QPVLNPEPNT VSYSQSSLIH LVGPSDCTLH GFVHGGVTMK LMDEVAGIVA ARHCKTNIVT ASVDAINFHD KIRKGCVITI SGRMTFTSNK SMEIEVLVDA DPVVDSSQKR YRAASAFFTY VSLSQEGRSL PVPQLVPETE DEKKRFEEGK GRYLQMKAKR QGHAEPQP (14-370 aa encoded by BC017365 )
This gene encodes a member of the acyl coenzyme family. The encoded protein hydrolyzes the CoA thioester of palmitoyl-CoA and other long-chain fatty acids. Decreased expression of this gene may be associated with mesial temporal lobe epilepsy. Alternatively spliced transcript variants encoding distinct isoforms with different subcellular locations have been characterized.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 50-kDa brain acyl-CoA hydrolase; ACH1; ACT; acyl-CoA thioesterase 2; Acyl-CoA thioesterase 7; acyl-CoA thioesterase, long chain; BACH; brain acyl CoA hydrolase; Brain acyl-CoA hydrolase; CTE-II; CTE-IIa; CTE-IIb; Cytosolic acyl coenzyme A thioester hydrolase; L RP1-120G22.10; LACH1; Long chain acyl-CoA thioester hydrolase; MTE-II
Gene Aliases: 2410041A17Rik; ACH1; ACOT7; ACT; BACH; CTE-II; CTE-IIa; hBACH; LACH; LACH1
UniProt ID: (Human) O00154, (Mouse) Q91V12, (Rat) Q64559
Entrez Gene ID: (Human) 11332, (Mouse) 70025, (Rat) 26759
Molecular Function:
esterase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support